Difco™ Bile Salts No. 3, BD Biosciences
Supplier: BD
Certificates
About this item
Bile Salts No
- Selective agents
- Bile extract
- Effective at less than 1/3 concentration of bile salts
Bile salts no. 3 contains bile extract standardized to provide inhibitory properties for selective media. It is a modified fraction of bile acid salts, providing a refined bile salt. Bile salts no. 3 is effective at less than one-third concentration of bile salts.
3 are used as selective agents for the isolation of gram-negative microorganisms, inhibiting gram-positive cocci.
Frequently Bought Together