À propos de cet article
The Mag-Bind® Plant DNA 96 Kit allows rapid and reliable isolation of high-quality genomic DNA from a wide variety of difficult to lyse and high polysaccharide plant species and tissues.
- Simple, rapid, and reliable procedure
- Reproducible yields of pure, ready to use DNA
- Adaptable to most robotic liquid handling platform
The system utilizes a CTAB based lysis buffer without the need for a chloroform or other organic solvent steps
The Mag-Bind® Plant DNA Kit also provides greater flexibility than silica plate based systems as partial plates can be used and can be scaled up to a 24-well format with the appropriate magnetic stand. Purified DNA is suitable for PCR, qPCR, restriction digestion, hybridization applications, and more demanding applications such as RAPD.
The propietary wash buffer systems eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. This kit is designed for manual or fully automated high throughput preparation of genomic, chloroplast, and mitochondrial DNA.
Spécifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Description:Mag-Bind® Plant DNA Plus 96 Kit