Glassware Washers Accessories
Glassware Washers Accessories
Supplier:   SP Industries
Glassware Washers Accessories
Supplier:  SP Industries
Certificates
Glassware Washers Accessories
22 Options Available Below
Product Details & Documents
Accessories designed for use with VWR® Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
  • Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
  • Bottom mesh inserts prevent small items from falling through
  • Made of stainless steel for long life and durability
  • Booster pump for use with low pressure DI water supply below 18 psi
See details in table below for specific use information. For further assistance, view the product literature or contact your Avantor representative.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 10002-914
  • 10002-904
  • 10002-944
  • 10002-908
  • 10002-932
  • 10002-934
  • 10002-936
  • 10002-938
  • 10002-928
  • 10002-930
  • 10002-942
  • 10002-940
  • 10002-906
  • 10002-912
  • 10002-924
  • 10002-910
  • 10002-916
  • 10002-918
  • 10002-920
  • 10002-922
  • 10002-926
  • 82021-092
  • Electrical:
  • 120v-1ph-60hz
  • For use with:
  • Half size rack baskets
  • Full size rack baskets
  • Low pressure DI water supply below 18psi
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    For Use With
    Fits
    Availability
    Price
  • Arrow clip rack insert, stainless steel, miscellaneous glassware on 14× small and 14× large arrow clips
    ½ of rack, bottom only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 631,95 $
    1 631,95 $
    Each
       1 631,95 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-914
    Electrical
    For use with
  • Basket and cover, stainless steel, general use (small glassware), 7.5L×7.5Wx4"D
    ¼ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 672,79 $
    672,79 $
    Prix promotionnel pour les nouveaux clients
    Each
       672,79 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-904
    Electrical
    For use with
  • Basket cover, stainless steel
    ½ size baskets (10002-908, 10002-910, 10002-912, 10002-924)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 451,36 $
    451,36 $
    Each
       451,36 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-944
    Electrical
    For use with
  • BOD bottle insert, stainless steel, for upto 18× BOD bottles
    ½ rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 681,14 $
    681,14 $
    Each
       681,14 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-908
    Electrical
    For use with
  • Bottle basket insert, stainless steel, 50 ml, for upto 69×50 ml bottles
    Full rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 854,60 $
    854,60 $
    Each
       854,60 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-932
    Electrical
    For use with
  • Bottle basket insert, stainless steel, 100 ml, for upto 40×100 ml bottles
    Full rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 043,00 $
    1 043,00 $
    Each
       1 043,00 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-934
    Electrical
    For use with
  • Bottle basket insert, 250 ml, stainless steel, for up to 24×250 ml bottles
    Full rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 043,25 $
    1 043,25 $
    Each
       1 043,25 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-936
    Electrical
    For use with
  • Bottle basket insert, 500 ml, stainless steel, for up to 23×500 ml bottles
    Full rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 862,27 $
    862,27 $
    Each
       862,27 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-938
    Electrical
    For use with
  • Bottle basket insert, stainless steel, 1 L, for upto 16×1 L bottles
    Full rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 252,15 $
    1 252,15 $
    Each
       1 252,15 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-928
    Electrical
    For use with
  • Bottle basket insert, stainless steel, 2 L, for upto 9×2 L bottles
    Full rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 782,69 $
    782,69 $
    Each
       782,69 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-930
    Electrical
    For use with
  • Bottom mesh insert, half size, stainless steel, tight mesh retains small items; prevents items from falling through basket
    ½ size baskets
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 094,65 $
    1 094,65 $
    Each
       1 094,65 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-942
    Electrical
    For use with
    Half size rack baskets
  • Bottom mesh insert, full size, stainless steel, tight mesh retains small items; prevents items from falling through basket
    Full size baskets
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 966,80 $
    966,80 $
    Each
       966,80 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-940
    Electrical
    For use with
    Full size rack baskets
  • Erlenmeyer flask insert, stainless steel, 300 - 1500 ml, for up to 5 × Erlenmeyer flasks
    ½ rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 910,83 $
    910,83 $
    Each
       910,83 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-906
    Electrical
    For use with
  • Graduated cylinder basket insert, stainless steel, for upto 32 small graduated cylinders
    ½ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 591,47 $
    591,47 $
    Each
       591,47 $
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-912
    Electrical
    For use with
  • Open basket insert with handles, stainless steel, general use (small glassware)
    ½ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 780,59 $
    1 780,59 $
    Each
       1 780,59 $
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-924
    Electrical
    For use with
  • Petri dish basket insert, stainless steel
    ½ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 333,31 $
    1 333,31 $
    Each
       1 333,31 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-910
    Electrical
    For use with
  • Test tube basket insert with lid, stainless steel, 75 mm tubes, for upto 150×75 mm long test tubes
    ½ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 896,33 $
    1 896,33 $
    Each
       1 896,33 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-916
    Electrical
    For use with
  • Test tube basket insert with lid, stainless steel, 105 mm tubes, for upto 150×105 mm long test tubes
    ½ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 356,50 $
    2 356,50 $
    Each
       2 356,50 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-918
    Electrical
    For use with
  • Test tube basket insert with lid, stainless steel, 165 mm tubes, for upto 150×165 mm long test tubes
    ½ of rack, bottom only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 920,14 $
    1 920,14 $
    Each
       1 920,14 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-920
    Electrical
    For use with
  • Test tube basket insert with lid, stainless steel, 200 mm tubes, for upto 150×200 mm long test tubes
    ½ of rack, bottom only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 945,39 $
    1 945,39 $
    Each
       1 945,39 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-922
    Electrical
    For use with
  • Watch glass insert, stainless steel, for upto 40 pieces less than 125 mm in diameter
    ½ of rack, top or bottom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 537,47 $
    537,47 $
    Each
       537,47 $
    Minimum order quantity is 1
    Spécifications:
    More
    Specifications
    Cat. No.
    10002-926
    Electrical
    For use with
  • Booster pump, 120 V / 1 phase / 60 Hz, for low pressure DI water supply below 18 psi
    Low pressure DI water supply below 18 psi
    VWR® glassware washers with DI rinse
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 265,18 $
    2 265,18 $
    Each
       2 265,18 $
    Spécifications:
    More
    Specifications
    Cat. No.
    82021-092
    Electrical
    120v-1ph-60hz
    For use with
    Low pressure DI water supply below 18psi