Bench Top Ionizer, Transforming Technologies | Avantor
Bench Top Ionizer, Transforming Technologies
IN5110 bench top ionizer, 7.1x2.7x7.5"
Catalog # 10837-124
Fournisseur #:  IN5110
Bench Top Ionizer, Transforming Technologies
Catalog # 10837-124
Bench Top Ionizer, Transforming Technologies
Bench Top Ionizer, Transforming Technologies
IN5110 bench top ionizer, 7.1x2.7x7.5"
Price
$975.00
Log in pour la disponibilité
Each
Product Details & Documents
The Model IN5110 68KHz AC benchtop air ionizer is a small, efficient static elimination device with variable fan speed for protecting electronic components and general static protection
A specialized piezoelectric high voltage transformer makes Ptec™ ionizers among the most reliable ionizers manufactured. Small, lightweight and stable, Ptec™ products are designed to remain in balance and to alarm when the HV output affects performance. The model IN5110 benchtop static eliminator produces a 68KHz AC output of approximately 2200V and a continuous stream of balanced air ions. Ionizers that use Ptec™ technology do not require calibration and only minimal maintenance.
Using AC corona technology, to make the air more conductive, Ptec™ ionizers produce a balanced stream of positive and negative air ions that never need calibration. These devices are used in many indus-tries and manufacturing applications where static electricity is a problem.
Spécifications
  • Désignation:
    IN5110 bench top ionizer
  • W×D×H:
    7.1x2.7x7.5"
  • Cat. No.:
    10837-124
  • Decay time:
    1000 - 100 V < 2 s at 1 ft.
  • Coverage area:
    1x5 ft.
  • Air volume:
    115 cfm, typical, high speed