PolarPack® Therapeutic Gel Packs, Flexible Gel Ice Packs, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
À propos de cet article
Cold and warm pack therapy used by physical therapists, hospitals, and physicians as a therapeutic compress to treat areas of pain and inflammation.
- Available in several sizes
- Can be used for either cold or warm compress
- Promotes faster healing
- Reusable
- Non-toxic
ThermoSafe Flexible PolarPacks® are a trusted solution to help promote injury recovery.
For warm compress, place the Flexible PolarPack® in warm water (do not exceed 150°F), or microwave on low setting. For cold compress, place in freezer until frozen. Use a cloth between the pack and the injured area. Do not exceed 20 minutes per application. 100% water soluble gel. Dispose with ordinary solid waste or dilute with water and dispose in conventional sink drain.
Frequently Bought Together