MetroMax i® Lab Carts, Metro
Fournisseur: Metropolitan Wire
Certificats
À propos de cet article
Lab carts with superior corrosion resistance.
All shelves adjust in 0.5" increments, offering precise shelf adjustment and maximum storage utilization.
Ordering information: All carts include 5MPX series casters (2 brake, 2 swivel), unless otherwise noted. Carts are not previously assembled.
Frequently Bought Together