Alcian Blue/PAS Special Stain Kit, Epredia™
Catalog # CA84000-254
Supplier: Thermo Scientific
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Periodic Acid Solution (0.5%)
- Cat. no.:CA84000-254
- Supplier No.:88016
Specifications
About this item
Demonstrate the presence of both neutral and acidic mucosubstances with this stain kit.
Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections. Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances.
Ordering information: Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent.