HEPA Filter, Labconco®
À propos de cet article
Replacement HEPA filter for use with XPert filtered systems and stations, XPert TXE equipment enclosures, FilterMate portable exhausters, Purifier logic biosafety sabinets, Purifier Cell Logic biosafety cabinets and Double HEPA filtration kits.
- 99.99% efficient on particles 0.3 µm in size
- Available in 18.0", 24.0", 30.0", and 42.0" widths
Have aluminum frames with either aluminum separators or separatorless pleats.
Ordering information: Note: Units 5' and 6' wide require two filters.
2 Options Available Below
Product Details & Documents
Replacement HEPA filter for use with XPert filtered systems and stations, XPert TXE equipment enclosures, FilterMate portable exhausters, Purifier logic biosafety sabinets, Purifier Cell Logic biosafety cabinets and Double HEPA filtration kits.
- 99.99% efficient on particles 0.3 µm in size
- Available in 18.0", 24.0", 30.0", and 42.0" widths
Have aluminum frames with either aluminum separators or separatorless pleats.
Ordering information: Note: Units 5' and 6' wide require two filters.
Product Family Options
Informations sur le produit
- DésignationFor Use WithHauteurLargeurWeightProfondeurAvailabilityPrice
- Replacement HEPA Filter4' XPert Systems, 4' XPert Stations, 4' Double HEPA Filtration Kits8.4 cm (3.3")106.7 cm (42.0")30 kg (66 lbs.)45.7 cm (18.0")Log in pour la disponibilitéEach$1,490.00Supplier Number: 3707902
- Supplier Number: 3838502
- Replacement Supply HEPA Filter for 5' Purifier LogicDiscontinued-Supplier Number: -