VWR® Contour™ ADA Height Base Cabinets, Fume Hood Cabinets
Supplier: VWR International
Certificates
About this item
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 83.8 cm (33"). All cupboard cabinets are supplied with one full-width adjustable shelf.
Frequently Bought Together