TE Buffers, Teknova
TE Buffers, Teknova
Supplier:   Teknova
TE Buffers, Teknova
Supplier:  Teknova
Certificates
TE Buffers, Teknova
8 Options Available Below
Product Details & Documents
For use in research and development.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Type de conditionnement
    Utilisation
    pH
    Availability
    Price
  • TE Buffer
    1 ml/tube
    50 tubes/rack
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $513.03
    $513.03
    Each
       $513.03
    Minimum order quantity is 1
    Supplier Number: T0224
  • TE Buffer
    250 ml
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (250ml)
    Each (250ml) - $239.43
    $239.43
    Each (250ml)
       $239.43
    Minimum order quantity is 1
    Supplier Number: T0225
  • TE Buffer
    1 L
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1,000ml)
    Each (1,000ml) - $316.88
    $316.88
    Each (1,000ml)
       $316.88
    Minimum order quantity is 1
    Supplier Number: T0226
  • TE Buffer, 5X
    1 L
    PCR
    8.0
    Discontinued
    Supplier Number: T3410
  • TE Buffer, 10X
    1 ml/tube
    50 tubes/rack
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Discontinued
    Supplier Number: T3455
  • TE buffer, 10X
    100 ml
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1ml)
    Each (1ml) - $266.80
    $266.80
    Each (1ml)
       $266.80
    Minimum order quantity is 10
    Supplier Number: T3456
  • TE Buffer, 10X
    250 ml
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100ml)
    Each (100ml) - $370.85
    $370.85
    Each (100ml)
       $370.85
    Minimum order quantity is 1
    Supplier Number: T3457
  • DNA Suspension Buffer
    250 ml
    PCR?, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (250ml)
    Each (250ml) - $129.76
    $129.76
    Each (250ml)
       $129.76
    Minimum order quantity is 1
    Supplier Number: T0223
  • DNA Suspension Buffer
    1 L
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1,000ml)
    Each (1,000ml) - $366.96
    $366.96
    Each (1,000ml)
       $366.96
    Minimum order quantity is 1
    Supplier Number: T0220
  • TE buffer, 10X
    100 ml
    PCR, Litigation, Restriction Digestion, DNA Sequencing
    8.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.19
    $103.19
    Each
       $103.19
    Minimum order quantity is 1
    Supplier Number: T3456
    Spécifications:
    New Product