AccuGENE™ Buffers
Supplier:   Lonza
AccuGENE™ Buffers
Supplier:  Lonza
Certificates
AccuGENE™ Buffers
2 Options Available Below
Product Details & Documents
AccuGENE® buffers are prepared with high quality reagents and use 18 megohm water
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
  • Formulation - 3.0 M NaCl, 0.3 M sodium citrate, pH 7.0
Store all reagents at 18 to 26 °C (64 to 79 °F).
They are manufactured according to strict quality control standards to ensure lot-to-lot consistency.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • CA12001-388
  • CA12001-394
  • Formulation:
  • 20X SSC with 3.0 M NaCl and 0.3 M Na3C6H5O7
  • 20X SSPE with 3.0 M NaCl, 0.2 M NaH2PO4 ò H2O, and 0.02 M EDTA
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    pH
    Availability
    Price
  • SSC buffer (20X)
    1 L
    7.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $90.50
    $90.50
    Each
       $90.50
    Supplier Number: 51205
    Spécifications:
    More
    Specifications
    Cat. No.
    CA12001-388
    Formulation
    20X SSC with 3.0 M NaCl and 0.3 M Na3C6H5O7
  • SSPE buffer (20X)
    1 L
    7.4
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $159.81
    $159.81
    Each
       $159.81
    Supplier Number: 51214
    Spécifications:
    More
    Specifications
    Cat. No.
    CA12001-394
    Formulation
    20X SSPE with 3.0 M NaCl, 0.2 M NaH2PO4 ò H2O, and 0.02 M EDTA