Zorbentz Wipes, Lint-Free, Cancer Diagnostics
Catalog # 10809-810
Supplier: CANCER DIAGNOSTICS, INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Zorbentz Wipes Lint-Free
- Dimensions:13 x 14 1/2"
- Cat. no.:10809-810
- Supplier No.:LP1111
Specifications
About this item
Zorbentz are a strong, highly absorbent, lint free alternative to paper towels and gauze.
- Ideal for coverslipping
- Prepare slides
- Soak up liquids and common solvents
- Individually folded
Zorbentz are a strong, highly absorbent, lint free alternative to paper towels and gauze.
Zorbentz cloth-like softness, with no loose threads or fibers, makes them ideal for use in coverslipping, slide preparation and general laboratory clean up.