Vapor Pad, Cancer Diagnostics
Catalog # 10809-806
Supplier: CANCER DIAGNOSTICS, INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Vapor Pad
- Dimensions:11 x 14"
- Cat. no.:10809-806
- Supplier No.:VP01114
Specifications
About this item
Formalin Neutralizer Pads encapsulate and neutralize harmful vapors to greatly reduce exposure to Formaldehyde.
- Offers contrast for all biopsies
- Cut resistant
- Durable
- Disposable
- Neutralizes harmful vapors
Formalin Neutralizer Pads that encapsulate and neutralize harmful vapors to greatly reduce exposure to Formaldehyde. Light blue contained work surface offers contrast for all biopsies. Cut resistant, durable and disposable.