ESD Nesting Container, MFG Tray
ESD-safe nesting boxes combine impressive strength, durability, and ease of handling with complete electrostatic discharge protection for sensitive parts
ESD-safe nesting box lids protect parts during storage and enable boxes to be stacked. ESD Lids combine impressive strength, durability, and ease of handling with complete electrostatic discharge protection for sensitive parts.
ESD-safe conductive products have a surface resistivity of less than 105 to 106 ohms. Fibrestat products meet or exceed ANSI/ESD S20.20 standard for ESD protection. And these boxes possess superior physical properties over thermoplastic and corrugated products. Nesting design saves valuable floor and bench space and facilitates the movement of boxes to work areas. Optional lids ensure complete product protection from dust and contamination, and enable boxes to be stacked. Fibrestat nesting boxes are resistant to cutting oils, solvents, greases, and mild acids and alkaline solutions within a pH rating of 3.0 to 10.0. Composite construction ensures high load-carrying capacity and dimensional stability.
ESD-safe nesting boxes combine impressive strength, durability, and ease of handling with complete electrostatic discharge protection for sensitive parts
ESD-safe nesting box lids protect parts during storage and enable boxes to be stacked. ESD Lids combine impressive strength, durability, and ease of handling with complete electrostatic discharge protection for sensitive parts.
ESD-safe conductive products have a surface resistivity of less than 105 to 106 ohms. Fibrestat products meet or exceed ANSI/ESD S20.20 standard for ESD protection. And these boxes possess superior physical properties over thermoplastic and corrugated products. Nesting design saves valuable floor and bench space and facilitates the movement of boxes to work areas. Optional lids ensure complete product protection from dust and contamination, and enable boxes to be stacked. Fibrestat nesting boxes are resistant to cutting oils, solvents, greases, and mild acids and alkaline solutions within a pH rating of 3.0 to 10.0. Composite construction ensures high load-carrying capacity and dimensional stability.
- Catalog No:
- 89022-962
- 89022-972
- 89022-970
- 89022-966
- 89022-974
- 89022-964
- 89022-968
- 89022-956
- 89022-960
- 89022-976
- pH range:
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- Temperature range:
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
Product Family Options
- DésignationDimensionsColor
- : 9221005167
Specifications
Cat. No.89022-962pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9261005167
Specifications
Cat. No.89022-972pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9241005167
Specifications
Cat. No.89022-970pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9231005167
Specifications
Cat. No.89022-966pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9301005167
Specifications
Cat. No.89022-974pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9221105167
Specifications
Cat. No.89022-964pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9231105167
Specifications
Cat. No.89022-968pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9201105167
Specifications
Cat. No.89022-956pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9211105167
Specifications
Cat. No.89022-960pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently - : 9301105167
Specifications
Cat. No.89022-976pH range3.0 to 10.0Temperature range–51 to 121°C (–60 to 250°F), up to 300°F intermittently