Nesting Container, MFG Tray | Avantor
Nesting Container, MFG Tray
Nesting Container, MFG Tray
Certificates
Nesting Container, MFG Tray
4 Options Available Below
Product Details & Documents
MFG Tray's durable nesting containers are resistant to cutting oils, grease, and solvents
Fiberglass lids will protect the contents of durable Nesting Containers.
Ordering Information: Comes in carton of 36.
Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for work-in-process part handling. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats, and totes for material handling in the metalworking, plastics, and electronics assembly industries, as well as in food service, confectionery, and pharmaceutical processing. MFG Tray products combine outstanding performance features with long service life to ensure exceptional value. The material matters in material handling.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89177-126
  • 89177-124
  • 89022-784
  • 89022-786
  • pH range:
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • Temperature range:
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Color
    Availability
    Price
  • Nesting Container
    9.75 x 6.13 x 4.5"
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $51.09
    $51.09
    Each
       $51.09
    Supplier Number: 9261085269
    Spécifications:
    More
    Specifications
    Cat. No.
    89177-126
    pH range
    3.0 to 10.0
    Temperature range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • Nesting Container
    9.75 x 9.25 x 2.13"
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $51.17
    $51.17
    Each
       $51.17
    Supplier Number: 9241085269
    Spécifications:
    More
    Specifications
    Cat. No.
    89177-124
    pH range
    3.0 to 10.0
    Temperature range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • Nesting Container
    11.4 x 9.25 x 3.4"
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $79.60
    $79.60
    Each
       $79.60
    Supplier Number: 9401085269
    Spécifications:
    More
    Specifications
    Cat. No.
    89022-784
    pH range
    3.0 to 10.0
    Temperature range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • Lid for 940108 series Nesting Container
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $64.58
    $64.58
    Each
       $64.58
    Supplier Number: 9401185269
    Spécifications:
    More
    Specifications
    Cat. No.
    89022-786
    pH range
    3.0 to 10.0
    Temperature range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently