ESD Conveyor/Assembly Tray, MFG Tray
Fournisseur: Molded Fiber Glass Tray
Certificats
À propos de cet article
Used in the assembly of circuit boards, computers, medical device, and other types of electronic or static sensitive assembly
Trays are designed for component assembly and for compatability with conveyors or cart transport systems.
Spécifications
- Catalog No:
- 89022-930
- 89022-934
- 89177-118
- 89022-944
- pH range:
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- 3.0 to 10.0
- Temperature range:
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently
- –51 to 121°C (–60 to 250°F), up to 300°F intermittently