Sklar's® Key Elevator is used in neurological procedures for lifting soft tissue and periosteum.
- Premium OR-Grade
It can also be used to lift bone in order to clear the surgery site. It features a solid, hexagonal handle with a gently curved blade tip with a sharp edge. It is available in several lengths with varying blade widths.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 10196-842
- 10196-884
- 10194-068
- 10196-850
- 10196-848
- 10196-846
- 10196-844
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free