To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Merit™ Bruns Plaster Shears
- Length:9½"
- Material:Stainless steel
- Cat. no.:10184-596
- Supplier No.:97-256
Specifications
About this item
Merit™ Bruns Plaster Shears are non-ratcheted finger ring specialty scissors designed for use with plaster and fiberglass casting
These scissors should never be used on tissue. They are heavy shears with one larger ring for the thumb. The lower blade has heavy serrations and a blunted probe end so as not to cause injury to the patient while removing casting materials. It is also slightly longer than the standard blade. The standard blade tip is also blunt. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product has slightly curved blades with a 20cm stop peg in between the finger rings. It has overall length of 9-1/2 inches, measured from the bottom of the larger finger ring to the tip of the serrated blade.