Carbon Film on Thin Bar Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
A thin film of pure carbon is deposited on the shiny side of the grid.
- Select copper, nickel, or gold grids in 200, 300, 4000, or 1000 mesh
- Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 - 6 nm), thick (15 - 20 nm), or extra thick (25 - 50 nm).
Packaging: Packed in grid storage box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together