Citric Acid Powder Pillows for Silica, 25 ml, Hach
Catalog # CA11016-958
Supplier: Hach
CAS Number:
Specifications
- Description:Citric Acid Powder Pillows for Silica
- Size:25 mL
- Method:Photometric
- Cat. no.:CA11016-958
- Supplier No.:1454828
- No. of Tests:1000
Specifications
About this item
For silica determination by photometric methods.
Packaging: Pack of 100 Powder Pillows.