The Deaver Retractor is a large, handheld retractor commonly used to hold back the abdominal wall during abdominal or thoracic procedures. It may also be used to move or hold organs away from the surgical site.
The Deaver retractor is shaped like a question mark with a thin, flat blade
Censitrac Ready products are industry standard and constructed of stainless steel.
It is available with a grip handle which is often held by a surgical assistant during procedures, or a flat handle so the retractor can be clamped into a position.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 10194-034
- 10197-310
- 10197-308
- 10197-306
- 10197-320
- 10197-732
- 10197-318
- 10197-316
- 10197-314
- 89025-402
- 10197-312
- 89025-400
- 10197-722
- 10197-724
- 10197-726
- 10197-728
- 10197-730
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free