À propos de cet article
Sklar's® Stille Rongeur is a double-action, debulking instrument used to scoop away bone that may itself be in need of manipulation or may be blocking access to a surgical site.
- Reusable and latex-free
- Non sterile
- Premium OR-grade
- Made of stainless steel
The plier handle has one straight arm on the bottom, while the other is slighlty curved with grooves and a horn thumb rest for increased grip and control. The double-action feature of this instrument allows greater force to be applied to the bone with less effort from the user.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Catalog No:
- 10195-828
- 10195-826
- Latex/Latex-free:
- Latex-free
- Latex-free