ContactUsLinkComponent
Instrument Cleaning Brush Set, Sklar®
Catalog # 10163-858
Supplier:  Sklar
undefined
Instrument Cleaning Brush Set, Sklar®
Catalog # 10163-858
Supplier:  Sklar
Supplier Number:  10-1426
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Bristle Material:
    Nylon
  • Handle Material:
    Stainless Steel/Plastic
  • Description:
    Instrument Cleaning Brush Set
  • Cat. no.:
    10163-858
  • Supplier No.:
    10-1426
Specifications
About this item
Instrument Cleaning Brush Set Include #10-1660 Cannula Cleaning Brush 12" 3mm 3/pk, #10-1362 Cannula Cleaning Brush 18" 5mm 3/pk, #10-1374 Cannula Cleaning Brush 24" 10mm 3/pk, #10-1376 Cannula Cleaning Brush 24" 10mm 3/pk, 10-1396 Suction Tube Brush 6" 1.6mm 3/pk, #10-1400 Suction Tube Brush 8" 2mm 3/pk, #10-1402 Suction Tube Brush 8" 2.6mm 3/pk, #10-1404 Suction Tube Brush 8" 3.3mm 3/pk, #10-1406 Suction Tube Brush 8" 4mm 3/pk, #10-1418 Surface Cleaning Brush 3/pk, #10-1444 Double End Brush Nylon 3/pk, #10-1650 Brush Stainless Steel Bristle 3/pk, #10-1651 Brush Nylon Bristle 3/pk - Set.