Quick Release Iris Hook, OR Grade, Sklar
À propos de cet article
The Sklar Iris Hook is a versatile instrument that can be used in many procedures to hold and retract tissue.
It is similar to, but smaller than the Emmett Hook, and the hook is slightly angled. The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length.
7 Options Available Below
Product Details & Documents
The Sklar Iris Hook is a versatile instrument that can be used in many procedures to hold and retract tissue.
It is similar to, but smaller than the Emmett Hook, and the hook is slightly angled. The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length.
Product Family Options
Informations sur le produit
- LongueurMatièreTip ConfigurationAvailabilityPrice
- Supplier Number: 90-8336
- Supplier Number: 90-8335
- Supplier Number: 90-8348
- Supplier Number: 90-8346
- Supplier Number: 90-8334
- 22.9 cm (9")Stainless steelQuick ReleaseLog in pour la disponibilitéLog in pour la disponibilitéPack of 4Pack of 4 - $1,579.96$1,579.96Prix promotionnel pour les nouveaux clientsPack of 4$1,579.96Supplier Number: 90-8332
- Supplier Number: 90-8344