Freer Elevator, OR Grade, Sklar®
Supplier:   Sklar
Freer Elevator, OR Grade, Sklar®
Supplier:  Sklar
Certificates
Freer Elevator, OR Grade, Sklar®
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar's® Freer Elevator is an orthopedic instrument that can be used as a multipurpose tool in many procedures, although it is primarily used for blunt debulking and lifting periosteum from bones in confined areas.
  • One tip is commonly blunt while the other is sharp
This double-ended instrument typically has a rounded handle with two slightly curved, tear-drop tips.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89024-976
  • 89024-978
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Latex/Latex-free:
  • Latex-free
  • Latex-free
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Grade
    Matière
    Curvature
    Sterility
    Tip Configuration
    Tip Dimensions
    Availability
    Price
  • Freer Elevator, Double End
    18.4 cm (7¼")
    Premium OR-Grade
    Stainless steel
    Slightly Angled
    Non sterile
    Sharp/Blunt
    5.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $199.49
    $199.49
    Prix promotionnel pour les nouveaux clients
    Each
       $199.49
    Supplier Number: 47-2380
    Spécifications:
    More
    Specifications
    Cat. No.
    89024-976
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
  • Freer Elevator, Double End
    19.7 cm (7¾")
    Premium OR-Grade
    Stainless steel
    Straight
    Non sterile
    Blunt/Sharp
    4.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $186.62
    $186.62
    Prix promotionnel pour les nouveaux clients
    Each
       $186.62
    Supplier Number: 47-2382
    Spécifications:
    More
    Specifications
    Cat. No.
    89024-978
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free