ContactUsLinkComponent
Econo™ Halsey Needle Holder, Floor Grade, Sklar®
Catalogue # 89023-412
Fournisseur:  Sklar
undefined
Econo™ Halsey Needle Holder, Floor Grade, Sklar®
Catalogue # 89023-412
Fournisseur:  Sklar
Supplier Number:  21-810
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    5"
  • Material:
    Stainless steel
  • Tip Configuration:
    Blunt
  • Curvature:
    Straight
  • Cat. no.:
    89023-412
  • Tip Style:
    Smooth
  • Supplier No.:
    21-810
Spécifications
À propos de cet article
Econo™ Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology.
Econo™ Halsey Needle Holders are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.