Prussian Blue Iron Stain Kit (Reaction for Demonstration of Iron), Polysciences
Catalog # 87001-921
Supplier: Polysciences Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Prussian Blue Iron Stain Kit
- Size:1 Kit
- Cat. no.:87001-921
- Supplier No.:24199-1
Specifications
About this item
Prussian Blue or Perls’ reaction is used to demonstrate ferric iron and ferritin
This is not a true staining technique, but rather a histochemical reaction. The protein is split off by the hydrochloric acid, allowing the potassium ferrocyanide to combine with the ferric iron. This forms the ferric ferrocyanide or Prussian Blue.