À propos de cet article
Sklar® Baron Suction Tube is an ENT instrument used for suctioning within narrow spaces, such as the ear canal.
Suction can be controlled by covering a small hole that is located on the thumb plate slightly above the handle. There is a slight angle to the instrument so that the user's hand does not obstruct the view of the area being suctioned. The Baron suction tube is available in French sizes 3, 5, or 7.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.