PYREX® Hybridization Tubes, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Hybridization Tubes, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Hybridization Tubes, Corning
2 Options Available Below
Product Details & Documents
PYREX® borosilicate glass, GL 45 orange plug-seal cap with silicone O-ring.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These tubes with three different lenghths are used in hybridiatzion incubators with rotator.
An optional Corning® vented membrane cap is available for low temperature assays.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 32645-030
  • 32645-032
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Ø×L
    Matière
    GL Thread Size
    Cap Type
    Availability
    Price
  • Hybridization Tubes, with Cap and O-Ring
    35×300 mm
    PYREX®
    45
    PP cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $310.00
    $310.00
    Case of 2
       $310.00
    Supplier Number: 7995-300
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    32645-030
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Hybridization Tubes, with Cap and O-Ring
    35×150 mm
    PYREX®
    46
    PP cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $281.00
    $281.00
    Case of 2
       $281.00
    Supplier Number: 7995-150
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    32645-032
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact