ContactUsLinkComponent
Corning® Direct Overhead Drive Center Cap Assembly for Corning® Reusable Spinner Flasks, Corning
Corning® Direct Overhead Drive Center Cap Assembly for Corning® Reusable Spinner Flasks, Corning
Catalog # 89089-676
Supplier:  Corning
Product available on GSA Advantage®
Corning® Direct Overhead Drive Center Cap Assembly for Corning® Reusable Spinner Flasks, Corning
Catalog # 89089-676
Supplier:  Corning
Supplier Number:  402614
Product available on GSA Advantage®
Specifications
  • Description:
    Direct overhead drive 100 mm center cap assembly
  • Cat. no.:
    89089-676
  • Supplier No.:
    402614
Specifications
About this item
The 120 mm Center Cap Assembly for Corning® Spinner Flasks is designed for use with direct overhead drive motor.
These cap assemblies required optional 8 L, 15 L or 36 L paddle assemblies and can be used on all 4512 series vertical sidearm flasks.