Corning® Polypropylene Bottles, Corning
 :   Corning
Corning® Polypropylene Bottles, Corning
 :  Corning
Corning® Polypropylene Bottles, Corning
Corning® Polypropylene Bottles, Corning
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
  • Hollow glass stopper style
  • Reusable
  • Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Product Family Options
  • Désignation
    Capacité
    Matière
    GL Thread Size
  • Narrow Mouth
    250 ml
    PP
    25
    Case of 12
    Case of 12 - $139.40
    $139.40
    Each
    Each - $139.40
    $139.40
    Case of 12
       $139.40
    Each
       $139.40
     : 1300P-250
  • Wide Mouth
    250 ml
    PP
    45
    Case of 12
    Case of 12 - $144.50
    $144.50
    Each
    Each - $144.50
    $144.50
    Case of 12
       $144.50
    Each
       $144.50
     : 1300P-250W
  • Narrow Mouth
    500 ml
    PP
    25
    Case of 12
    Case of 12 - $173.40
    $173.40
    Each
    Each - $173.40
    $173.40
    Case of 12
       $173.40
    Each
       $173.40
     : 1300P-500
  • Wide Mouth
    500 ml
    PP
    45
    Case of 12
    Case of 12 - $179.35
    $179.35
    Each
    Each - $179.35
    $179.35
    Case of 12
       $179.35
    Each
       $179.35
     : 1300P-500W
  • Narrow Mouth
    1000 ml
    PP
    32
    Case of 12
    Case of 12 - $251.60
    $251.60
    Each
    Each - $251.60
    $251.60
    Case of 12
       $251.60
    Each
       $251.60
     : 1300P-1L
  • Wide Mouth
    1000 ml
    PP
    63
    Case of 12
    Case of 12 - $266.05
    $266.05
    Each
    Each - $266.05
    $266.05
    Case of 12
       $266.05
    Each
       $266.05
     : 1300P-1LW