PYREX® Graham Condenser with Single [ST] Outer Top Joint, Corning
Fournisseur: Corning
Certificats
À propos de cet article
Condensers made of PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These Graham condensers have a high surface area per unit with a standard taper outer joint at the top and a plain drip tip at the bottom. They are primarily used in distillation assemblies. Condenser tubulations are approximately 10 mm O.D.
Spécifications
- Catalog No:
- 23138-029
- 23138-040
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey