PYREX® Trypsinizing Flasks with Baffles, Corning
2000 ml, PYREX® Trypsinizing Flasks with Baffles
Catalog # 22877-184
Supplier: Corning
Supplier #: 4460-2L
Jump to:
Product Details & Documents
Flasks are used for converting homogenous tissue samples into cell suspension by digestion of connective proteins
Heavy beaded neck accepts cotton plugs. Four extra-deep baffles enhance vigorous agitation.
Specifications
- Volume:2000 ml
- Désignation:PYREX® Trypsinizing Flasks with Baffles