PYREX® Conical Centrifuge Tube with Beaded Rim, Corning
: Corning
Made of PYREX® borosilicate glass, conical, beaded.
- Wide neck
- Excellent chemical resistance
- Excellent temperature resistance
These centrifuge tubes has uniform glass distribution in the wall, taper and bottom provides improved mechanical strength, prevents the formation of failure-causing stress regions during centrifuging and insures the highest degree of accuracy.
- Catalog No:
- 21021-202
- 21021-188
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Frequently Bought Together