PYREX® EZ Access™ Extra-Wide Neck Volumetric Flask, Class A, Corning
Supplier: Corning
Certificates
About this item
These PYREX® EZ Access™ heavy walled volumetric flasks are designed to meet Class A, ASTM tolerances for volumetrics.
- Calibrated 'to contain' (TC)
- GL 32 high temperature screw cap fits all flasks
- Screw cap with PTFE liner provides a secure closure for leak proof storage of solutions
- Extra large volume size is permanently screened onto the flask body for easy volume identification
The wide neck design allows for easy filling and mixing of various materials into solution and also allows access to larger capacity pipettes.
Specifications
- Catalog No:
- 66025-668
- 66025-666
- 66025-664
- 66025-662
- 66025-660
- 66025-670
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey