Corning® Polypropylene Closures, Corning
Supplier: Corning
Certificates
About this item
These Corning® polypropylene closures are designed for culture tubes and flasks.
- Internal drip ring minimizes contamination
- Autoclavable at 121 °C
Closure offers two positions: open for gas exchange or closed for humidified environment.
Frequently Bought Together