ContactUsLinkComponent
McBurney Retractor, OR Grade, Sklar
Catalogue # 10197-696
Fournisseur:  Sklar
undefined
McBurney Retractor, OR Grade, Sklar
Catalogue # 10197-696
Fournisseur:  Sklar
Supplier Number:  57-1475
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    McBurney Retractor
  • Length:
    19.1 cm (7½")
  • Material:
    Stainless steel
  • Tip Configuration:
    Fenestrated Blade
  • Cat. no.:
    10197-696
  • Supplier No.:
    57-1475
Spécifications
À propos de cet article
McBurney Retractors are constructed of stainless steel
Industry standard.