Glass Vials, C&G Containers
Glass Vials, C&G Containers
Glass Vials, C&G Containers
Supplier:  C&G Containers
Certificates
Glass Vials, C&G Containers
5 Options Available Below
Product Details & Documents
These glass vials from C&G Containers are designed primarily for the collection and testing of solid samples for Volatile Organic Analysis and Total Petroleum Hydrocarbons (TPH), and for the collection and testing of aqueous samples for Total Organic Carbon (TOC).
  • All glass vials are borosilicate Type 1 glass and use a 24-414 closure
Pre-cleaned and Certified products contain a Certificate of Analysis and Cleaning Protocol Form in the product case. Wash protocols for these vials include; A1 (Nitric acid wash) certified, B1 process certified, L3 (no wash, vial with cap attached) not certified, and D1 protocol, a depyrognation-like process for low level Total Organic Carbon (TOC), certified. Some vials are tare weighed. Some vials have dust covers.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Caps can be either one-piece Open Top polypropylene with PTFE septa ultrasonically bonded (not glued), two-piece Open Top polypropylene with pressure fitted PTFE septa, solid top polypropylene cap with PTFE septa, or solid top polypropylene cap with PTFE liner. The PTFE septa is a 3.1 mm fluoropolymer resin/silicone septa laminate (overall thickness 0.125", ±0.006", silicone thickness 0.115", PTFE 0.010"). There is also a thin bonded PTFE Septa cap, overall thickness 0.060 ±0.005" (silicone: 0.056", PTFE: 0.004"). The PTFE liner is a 0.10" thick fluoropolymer disc.
Secondary uses include containment of extracts. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity "contain" volumes may vary.
Documents
Product Family Options
Clear Glass Vials without Preservative
Informations sur le produit
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Cap Type
    Cap Color
    Septum
    Availability
    Price
  • Vial with sample label, B1 process protocol
    20.00 ml
    27.5×57 mm
    Clear
    Oui
    Open top, 1-Piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    20.00 ml
    27.5×57 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    20.00 ml
    27.5×57 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    20.00 ml
    27.5×57 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    20.00 ml
    27.5×57 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    20.00 ml
    27.5×57 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 2-piece
    White
    PTFE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - 201,69 $
    201,69 $
    Prix promotionnel pour les nouveaux clients
    Case of 72
       201,69 $
    Supplier Number: LV73A000A01BZS
  • Vial, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-Piece
    White
    PTFE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - 423,69 $
    423,69 $
    Prix promotionnel pour les nouveaux clients
    Case of 100
       423,69 $
    Supplier Number: LFV0403000
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    Thin PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-Piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    60.00 ml
    27.5×140 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
Amber Glass Vials without Preservative
Informations sur le produit
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Cap Type
    Cap Color
    Septum
    Availability
    Price
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, B1 process protocol
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE-lined
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - 173,38 $
    173,38 $
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - 173,38 $
    173,38 $
    Prix promotionnel pour les nouveaux clients
    Case of 80
       173,38 $
    Each
       173,38 $
    Supplier Number: LU23A000A01FZS
  • Vial, L3 process protocol (assembled only)
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    20.00 ml
    27.5×57 mm
    Amber
    Oui
    Solid top
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    Thin PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, Total Organic Carbon
    40.00 ml
    27.5×95 mm
    Amber
    Oui
    Open top, 1-piece
    White
    Thin teflon/silicon
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 80
    Case of 80 - 646,52 $
    646,52 $
    Prix promotionnel pour les nouveaux clients
    Case of 80
       646,52 $
    Supplier Number: LAV040700005
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial with sample label, B1 process protocol
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Vial, L3 process protocol (assembled only)
    60.00 ml
    27.5×140 mm
    Amber
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
Clear Glass Vials with Preservative
Informations sur le produit
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Cap Type
    Cap Color
    Septum
    Availability
    Price
  • Barcoded vial, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - 233,54 $
    233,54 $
    Prix promotionnel pour les nouveaux clients
    Case of 72
       233,54 $
    Supplier Number: LV74H100B21BZS
  • Barcoded vial, tare weighed with stirbar, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial, tare weighed with stirbar, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -
  • Barcoded vial with sample label, B1 process protocol
    40.00 ml
    27.5×95 mm
    Clear
    Oui
    Open top, 1-piece
    White
    PTFE
    Discontinued
    Supplier Number: -