ContactUsLinkComponent
Bailey Foreign Body Remover, OR Grade, Sklar
Bailey Foreign Body Remover, OR Grade, Sklar
Catalog # 10197-388
Supplier:  Sklar
undefined
Bailey Foreign Body Remover, OR Grade, Sklar
Catalog # 10197-388
Supplier:  Sklar
Supplier Number:  65-4080
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Bailey Foreign Body Remover
  • Material:
    Stainless steel
  • Tip Configuration:
    LOOP
  • Cat. no.:
    10197-388
  • Supplier No.:
    65-4080
Specifications
About this item
Sklar's Bailey Foreign Body Remover utilizes a small, latex-free plastic loop to expel foreign objects.
The main part of the remover (21/4" in length) is found inside a stainless steel sleeve that also acts as a handle extension, enabling the overall length to be extended to 43/4". The instrument may be used with or without the extender, depending on user preference.