Sodium hydroxyde solution standard 50%
500 ml
Catalog # CA11027-990
Supplier: Hach
Supplier #: 218049
Jump to:
Product Details & Documents
Multi-purpose sodium hydroxide solution.
Synonyms: Hydroxyde de sodium
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- UN:1824
- ADR:8,II
Specifications
Specifications
- Taille:500 ml



