HxSil C18 HPLC Guard Columns, Reversed Phase, Hamilton
Supplier: Hamilton
Certificates
About this item
Hamilton guard columns protect analytical, semi-prep and preparative HPLC columns.
- Protect columns
- Remove particulates
- Modular design
- Reduces dead volume
- Prevents column band broadening
Cartridge holders for analytical columns are available in stainless steel and PEEK materials. Semi-prep/preparative holders are available only in stainless steel. The design of the cartridge holders is optimized to reduce dead volume and prevent extra column band broadening. Cartridge holders are reusable indefinitely.
They also remove particulate contaminants and highly adsorptive compounds from samples, prolonging column life. The designs are modular and cartridge replacement is easy and tool-free.
Specifications
- Catalog No:
- 10836-974
- 10836-972
- 10837-060
- 10836-976
- 10837-058
- 10836-944
- 10836-966
- 10836-948
- 10836-968
- 10836-946
- 10836-978
- Description:
- 10836-974
- 10836-972
- 10837-060
- 10836-976
- 10837-058
- 10836-944
- 10836-966
- 10836-948
- 10836-968
- 10836-946
- 10836-978
- Application:
- Organics
- Organics
- Organics
- Organics
- Peptides/Proteins
- Organics
- Peptides/Proteins
- Peptides/Proteins
- Peptides/Proteins
- Organics
- Peptides/Proteins