À propos de cet article
Hamilton introduces a new headspace syringe featuring a unique Glue-Free (GF) needle attachment in combination with the well-known High Dynamic (HD) plunger designed specifically for PAL COMBI-xt Headspace Autosamplers.
- Glue-free needle
- High dynamic (HD) plunger
- Chemically inert
Glue-Free needle attachment is chemically inert, eliminating detached needles due to contact with organic, and chlorinated solvents. Glue-Free needle attachment minimizes ghost peaks. Temperature stability up to 200°C means a wider range of sample components can be analyzed. Patented spring-in-plunger design creates a dynamic seal between the plunger tip and the inside of the glass barrel for leak-free operation. Hamilton offers a wide range of quality syringe options for the PAL GC and Combi PAL (Headspace) GC Autosamplers.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10837-054
- 10836-942
- 75778-158
- 10836-940
- 10836-938
- 10836-936
- Min. temperature:
- 392°F (200°C)
- 392°F (200°C)
- 392°F (200°C)
- 392°F (200°C)
- 392°F (200°C)
- Needle length:
- 2"
- 2"
- 2"
- 2"
- 2"
- Needle style:
- 5
- 5
- 5
- 5
- 5