The Slide-A-Lyzer® G2 Dialysis Cassette offers efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.
- Pipette-accessible for easy sample loading and retrieval
- Self-floating chambers for buoyancy and vertical orientation during dialysis
- Designed to maintain the highest possible sample integrity and protection with rigorous quality testing for maximum consistency
- Fast and consistent dialysis with maximum sample recovery
- Ideal for removing low-molecular weight contaminants, performing buffer exchange and desalting
- Store at room temperature
The cassette membrane is composed of low-binding regenerated cellulose for maximum sample recovery while maintaining maximum sample purity and are manufactured using clean-room conditions to ensure cassettes are contaminant-free.
Volume range using a pipette: 15 to 30 ml
Volume range using a syringe: 10 to 30 ml
They are available in four precise membrane MWCOs (2K, 3.5K, 10K and 20K) for dialysing sample volumes up to 30 ml.