ContactUsLinkComponent
Fungi-Fluor® Kit for Fungal Detection, Polysciences
Catalog # 10819-090
Supplier:  Polysciences Inc
CAS Number:  
undefined
Fungi-Fluor® Kit for Fungal Detection, Polysciences
Catalog # 10819-090
Supplier:  Polysciences Inc
Supplier Number:  17442-1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fungi-Fluor® Kit for Fungal Detection
  • Cat. no.:
    10819-090
  • Supplier No.:
    17442
Specifications
About this item
The Fungi-Fluor® Kit for Fungal Detection offers a quick fluorescent stain/counterstain procedure for various fungal organisms
Kit contains 75mL of staining solution A, and 75mL of counterstaining solution B.
The kit can be used to screen a variety of specimen types, such as sputum and skin scrapings, for fungal detection.