SmartCal™ Moisture Analyzer Test Substance | Avantor
SmartCal™ Moisture Analyzer Test Substance
SmartCal™ Moisture Analyzer Test Substance
Supplier:  Mettler Toledo
Certificates
SmartCal™ Moisture Analyzer Test Substance
SmartCal™ Moisture Analyzer Test Substance
6 Options Available Below
Product Details & Documents
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
  • Simply pour SmartCal™ into the sample pan and start
  • Test takes just 10 minutes
  • Easy to monitor and interpret instrument's performance over time
  • Store results directly in the instrument or manually enter into a free validated Excel® report sheet
  • Done just like a normal moisture measurement
  • Certified version available to satisfy regulatory requirements
  • SmartCal™ to get you up and running right away
SmartCal™ is a granular test substance. Its high sensitivity reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices.
Testing regularly with SmartCal™ ensures that moisture measurements are reliable at all times.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Moisture analyzer test system, certified version (set of 12)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,450.00
    $1,450.00
    Each
       $1,450.00
    Supplier Number: 30005793
  • Moisture analyzer test system, certified version (set of 24)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,800.00
    $1,800.00
    Each
       $1,800.00
    Supplier Number: 30005791
  • Moisture analyzer test system (set of 12)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $994.00
    $994.00
    Each
       $994.00
    Supplier Number: 30005792
  • Moisture analyzer test system (set of 24)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,280.00
    $1,280.00
    Each
       $1,280.00
    Supplier Number: 30005790
  • Moisture analyzer test system starter pack, certified version (set of 12)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,150.00
    $1,150.00
    Each
       $1,150.00
    Supplier Number: 30005918
  • Moisture analyzer test system starter pack (set of 12)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $860.00
    $860.00
    Each
       $860.00
    Supplier Number: 30005917