Anti-PRNP Mouse Monoclonal Antibody [clone: 3F4]
Supplier: Biolegend
Certificates
About this item
Type: Primary
Antigen: PRNP
Clonality: Monoclonal
Clone: 3F4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Hamster, Feline
Frequently Bought Together