Anti-CD196 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: G034E3]
Supplier: Biolegend
Certificates
About this item
Alexa Fluor® 647 anti-human CD196 (CCR6) [G034E3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD196
Clonality: Monoclonal
Clone: G034E3
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Specifications
- Catalog No:
- CA353404-BL
- CA353403-BL
- Antigen symbol:
- CD196
- CD196
- Antigen synonyms:
- STRL22|GPRCY4|CCR6|GPR-CY4|CKR-L3|CKRL3|CMKBR6|CKR6|CD196|DRY-6|GPR29|BN-1|DCR2
- STRL22|GPRCY4|CCR6|GPR-CY4|CKR-L3|CKRL3|CMKBR6|CKR6|CD196|DRY-6|GPR29|BN-1|DCR2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Clone:
- G034E3
- G034E3
- Reactivity:
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1235
- 1235
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Immunogen:
- CCR6-transfected cells
- CCR6-transfected cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT