Roll-O-Matic® Self-Wringing Cleanroom Sponge Mops, Vileda Professional-FHP
Fournisseur: Vileda Professional - FHP
Certificats
À propos de cet article
The ideal self-wringing cleanroom mop
- Straight handles allow user to properly use the pull-and-lift technique with both sides of the mop
- Perfect for use on floors, walls, and ceilings
- Ergonomic grips reduce worker fatigue
- Entire aluminum and steel mop handles are completely autoclavable at the most common autoclave cycles
- Caps on steel versions are stainless steel and completely sealed
- Aluminum version features color coding tabs (blue, red, green, yellow) for easy equipment identification
The Vileda Professional® stainless steel and aluminum extendable Roll-O-Matic® mops are the best choice when seeking one mop to apply disinfectant, clean surfaces, or absorb spills in all production and controlled environments. These stainless steel Roll-O-Matic® mops are manufactured to resist repeated autoclave cycles and continued exposure to the common chemicals and disinfectants used in the controlled environment marketplace.
Frequently Bought Together