ContactUsLinkComponent
GCxGC Selectivity Kit (0.18 mm), Restek
Catalogue # 10850-640
Fournisseur:  Restek
undefined
GCxGC Selectivity Kit (0.18 mm), Restek
Catalogue # 10850-640
Fournisseur:  Restek
Supplier Number:  15130
Spécifications
  • Description:
    GCxGC (0.18 mm) Selectivity Kit
  • Cat. no.:
    10850-640
  • Packing Material:
    Kit0, e
  • Includes:
    Rxi-1ms column (cat.# 15120), Rxi-5Sil MS column (cat.# 15119), Rxi-XLB column (cat.# 15121), Rxi-17Sil MS column (cat.# 15116), Rtx-200 column (cat.# 15117), Stabilwax column (cat.# 15118), and a 5-pk. of deactivated universal press-tight connectors (cat.# 20429)
  • Supplier No.:
    15130
Spécifications
À propos de cet article
GCxGC selectivity kits offer orthogonal separations through a wide range of stationary phases and accommodate varying sample capacities, speeds, and detectors.
  • Wide range of stationary phases offers orthogonal separations
  • High thermal stability increases system ruggedness
  • Unrivaled column inertness for accurate analysis of active compounds
  • 0.15, 0.18, or 0.25 mm Int.Ø formats accommodate varying sample capacities, speeds, and detectors
  • Secondary columns come in convenient 2 m lengths for economical methods development
Selectivity kits include secondary columns that come in convenient lengths for economical development of methods.