ContactUsLinkComponent
MXT®-Alumina BOND/MAPD Columns (Siltek®-treated stainless steel PLOT), Restek
Fournisseur:  Restek
Certificats
undefined
MXT®-Alumina BOND/MAPD Columns (Siltek®-treated stainless steel PLOT), Restek
Fournisseur:  Restek
Spécifications
  • Catalog No:
  • 10851-168
  • 10851-412
  • Description:
  • MXT-Alumina BOND/MAPD, 3.5" Coil Configuration
  • MXT-Alumina BOND/MAPD, 7" 11-pin cage Configuration
  • Temperature limit:
  • to 250 °C
  • to 250 °C